Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUMA Antibody, Novus Biologicals™
SDP

Catalog No. NB500261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500261 0.1 mL
NB500261SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500261 Supplier Novus Biologicals Supplier No. NB500261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

PUMA Polyclonal specifically detects PUMA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PUMA
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 5.65 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:20-1:100
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. Q9BXH1
Gene Alias BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
Gene Symbols BBC3
Host Species Rabbit
Immunogen A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProt# Q9BXH1]
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 27113
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.