Learn More
Invitrogen™ PureLink™ Pro 96 PCR Purification Kit
Description
Includes
Four 96-well plates and sufficient reagents to perform four 96-well purifications
The PureLink™ Pro 96 PCR Purification Kit combines advanced silica plate extraction chemistry with an optimized 96-well plate design for manual or automated processing of PCR cleanup in as little as 20 minutes. The PureLink™ Pro 96 PCR Purification Kits provide rapid and efficient removal of short primers, dNTPs, enzymes, short-failed PCR products, and salts from PCR products. The simple PureLink™ bind/wash/elute procedure facilitates routine purifications of double-stranded DNA fragments from 100bp to 12kb.
Improved plate design
The PureLink™ Pro 96 PCR Purification Kit provides optimized semi-skirted 96-well plates for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. In addition, sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation, or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.
PCR products purified using the PureLink™ Pro 96 PCR Purification Kit demonstrate outstanding performance in downstream applications such as cloning automated fluorescent DNA sequencing, PCR, restriction mapping, and labeling.
High Throughput-Compatible
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | The PureLink™ Pro 96 PCR Purification Kit includes 4 x 96-well plates, and sufficient reagents to perform 4 x 96-well purifications. Store components at room temperature. All reagents are guaranteed stable for 12 months when properly stored. |
| Format | Kit |
| Elution Volume Max | 50 μL (centrifuge), 100 μL (vacuum), 150 μL |
| For Use With (Equipment) | Biomek™ FX, Robotic Liquid Handling Systems, Vacuum Manifold, Eppendorf epMotion, Centrifuge, TECAN Freedom Evo, TECAN Genesis |
| High-throughput Compatibility | High-throughput Compatible |
| Product Line | PureLink |
| Product Type | PCR Purification Kit |
| Purity or Quality Grade | Molecular Biology Grade |
| Quantity | 4 plates kit |
| Run Time | 20 min. |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.