Learn More
Purine Nucleoside Phosphorylase/PNP Antibody (103), DyLight 488, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Purine Nucleoside Phosphorylase/PNP |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 103 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.4.2.1, FLJ94043, FLJ97288, Inosine phosphorylase, MGC117396, MGC125915, MGC125916, NPFLJ97312, nucleoside phosphorylase, PRO1837, PUNP, purine nucleoside phosphorylase, purine-nucleoside:orthophosphate ribosyltransferase |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Purine Nucleoside Phosphorylase/PNP (P00491; Met 1-Ser 289). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.