Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purine Nucleoside Phosphorylase/PNP Rabbit anti-Human, Clone: 103, Novus Biologicals™
SDP

Catalog No. NB021570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021570 50 μg
NB021569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021570 Supplier Novus Biologicals Supplier No. NBP29005350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Purine Nucleoside Phosphorylase/PNP Monoclonal specifically detects Purine Nucleoside Phosphorylase/PNP in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Purine Nucleoside Phosphorylase/PNP
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 103
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias EC 2.4.2.1, FLJ94043, FLJ97288, Inosine phosphorylase, MGC117396, MGC125915, MGC125916, NPFLJ97312, nucleoside phosphorylase, PRO1837, PUNP, purine nucleoside phosphorylase, purine-nucleoside:orthophosphate ribosyltransferase
Host Species Rabbit
Immunogen Recombinant Human Purine Nucleoside Phosphorylase/PNP protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4860
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.