Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ ESK™ Sampling Kits with Pre-filled Buffered Peptone Water
Click to view available options
Volume (Metric):
4 mL
10 mL
Quantity:
300 Cs.
50 Cs.
Description
For environmental surface sampling; Intended as a nonselective media for the pre-enrichment of Salmonella spp. from food sources; Easy edge-peel packaging; Quick-turn leak-resistant cap; High-quality Puritan polyester tipped swab; Swab is securely attached to cap; Pre-filled with 4ml Buffered Peptone Water; Available in bulk case pack; Shelf life - 18 months; Made in USA
Specifications
Specifications
| Description | Sampling Kit |
| Material | Plastic |
| Sterility | Sterile |
| Volume (Metric) | 4 mL |
| For Use With (Application) | Environmental surface sampling with buffered peptone water and detection of microorganisms |
| Height (English) | 4 in. |
| Includes | 4mL or 10mL Butterfield's solution, 0.1% peptone and 2% Tween 80 |
| Volume (English) | 0.13 oz. |
| Quantity | 50 Cs. |
| Certifications/Compliance | ISO 9001:2008, ISO 13485:2003 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction