Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ ESK™ Sampling Kits with Pre-filled Letheen Broth
Click to view available options
Volume (Metric):
4 mL
10 mL
Quantity:
300 Cs.
50 Cs.
Description
Intended to inactivate quaternary ammonium compounds to determine the number of bacteria present in cosmetics and other materials; For environmental surface sampling; Easy edge-peel packaging; Quick-turn leak-resistant cap ;High-quality Puritan polyester tipped swab; Swab is securely attached to cap; Pre-filled with 4ml Letheen Broth; Available in bulk case pack; Shelf life - 18 months; Made in USA
Specifications
Specifications
Description | Sampling Kit |
Material | Plastic |
Sterility | Sterile |
Volume (Metric) | 4 mL |
For Use With (Application) | Environmental surface sampling with letheen broth and detection of microorganisms |
Height (English) | 4 in. |
Includes | Prefilled letheen broth |
Volume (English) | 0.13 oz. |
Height (Metric) | 103 cm |
Quantity | 50 Cs. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction