Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin-sensitive aminopeptidase/NPEPPS Antibody, Novus Biologicals™
SDP

Catalog No. NB10093421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093421 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093421 Supplier Novus Biologicals Supplier No. NB10093421
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Puromycin-sensitive aminopeptidase/NPEPPS Polyclonal specifically detects Puromycin-sensitive aminopeptidase/NPEPPS in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen Puromycin-sensitive aminopeptidase/NPEPPS
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.2 - 0.5 ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006301.2
Gene Alias AAP-S, aminopeptidase puromycin sensitive, Cytosol alanyl aminopeptidase, EC 3.4.11.2, EC 3.4.11.3, metalloproteinase MP100, MP100EC 3.4.11, PSAEC 3.4.11.14, puromycin-sensitive aminopeptidase
Gene Symbols NPEPPS
Host Species Goat
Immunogen Peptide with sequence C-KRDAESIHQYLLQRK corresponding to internal region according to NP_006301.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9520
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse and Rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.