Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puromycin-sensitive aminopeptidase/NPEPPS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Puromycin-sensitive aminopeptidase/NPEPPS |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Puromycin-sensitive aminopeptidase/NPEPPS Polyclonal specifically detects Puromycin-sensitive aminopeptidase/NPEPPS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Puromycin-sensitive aminopeptidase/NPEPPS | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9520 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: VMNCADIDIITASYAPEGDEEIHATGFNYQNEDEKVTLSFPSTLQTGTGTLKIDFVGELNDKMKGFYRSKYTTPSGEVRYAAVTQFEA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
AAP-S, aminopeptidase puromycin sensitive, Cytosol alanyl aminopeptidase, EC 3.4.11.2, EC 3.4.11.3, metalloproteinase MP100, MP100EC 3.4.11, PSAEC 3.4.11.14, puromycin-sensitive aminopeptidase | |
NPEPPS | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title