Learn More
MilliporeSigma Purospher™ STAR RP-18 Endcapped (2 μm) Capillary Column, 0.3 mm I.D.
Shop All MilliporeSigma Supelco ProductsDescription
- Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, catecholamines, explosives, oligonucleotides, esters, fat soluble vitamins, lipids, fatty acids, flavonoids, ketones, nitrosamines, nucleosides, PAHs, PCBs, peptides, pesticides, phenols, phthalates, preservatives, steroids, metabolized steroids, sulfonamides, sweeteners, and water soluble vitamins.
Specifications
Specifications
| Diameter (Metric) | 0.3 mm |
| Length (Metric) | 150 mm |
| Particle Size | 2 μm |
| Pore Size | 120 Å |
| Phase | Reversed Phase |
| Material | Stainless Steel |
| Technique | UHPLC |
| Product Type | UHPLC Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.