Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUS10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156369
Description
PUS10 Polyclonal specifically detects PUS10 in Human samples. It is validated for Western Blot.Specifications
PUS10 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q3MIT2 | |
PUS10 | |
Synthetic peptides corresponding to PUS10 (pseudouridylate synthase 10) The peptide sequence was selected from the middle region of PUS10. Peptide sequence AVFVAGRYNKYSRNLPQTPWIIDGERKLESSVEELISDHLLAVFKAESFN. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Xenopus: 100%; Bovine: 100%; Chicken: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Guinea pig: 92%; Zebrafish: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CCDC139, coiled-coil domain containing 139, Coiled-coil domain-containing protein 139, DOBIMGC126729, EC 5.4.99, EC 5.4.99.-, FLJ32312, MGC126755, pseudouridine synthase 10, pseudouridylate synthase 10, Psi55 synthase, PUS1, putative tRNA pseudouridine synthase Pus10, tRNA pseudouridine 55 synthase, tRNA pseudouridylate synthase, tRNA-uridine isomerase | |
Rabbit | |
Affinity Purified | |
RUO | |
150962 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction