Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Putative Multidrug Resistance Protein 1 Mouse anti-Plant, Clone: 1-6, Novus Biologicals™
SDP

Catalog No. NB036744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036744 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036744 Supplier Novus Biologicals Supplier No. NBP289161

Mouse Monoclonal Antibody

Putative Multidrug Resistance Protein 1 Monoclonal specifically detects Putative Multidrug Resistance Protein 1 in Plant samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Putative Multidrug Resistance Protein 1
Applications Immunofluorescence
Classification Monoclonal
Clone 1-6
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias ABC transporter B family member 11; ABC transporter B family member 9, mdr6, MDR-like ABC transporter, OJ1029_F04.19, Os01g0533900, putative multidrug resistance protein 1
Host Species Mouse
Immunogen Recombinant protein within rice Putative Multidrug Resistance Protein 1 aa 427-595/ 843. (SwissProt: Q5JLI4 Rice)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4326410
Target Species Plant
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.