Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puumala Virus Glycoprotein Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB201684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201684 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201684 Supplier Novus Biologicals Supplier No. NBP241260H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Puumala Virus Glycoprotein Polyclonal antibody specifically detects Puumala Virus Glycoprotein in Virus samples. It is validated for ELISA

Specifications

Antigen Puumala Virus Glycoprotein
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Envelopment polyprotein, Gc, Glycoprotein C, Glycoprotein G1, Glycoprotein G2, Glycoprotein N, Gn, M polyprotein, Precursor, PUUVsMgp1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the amino terminus of the Puumala virus glycoprotein. The immunogen is located within amino acids 90 - 140 of Puumala Virus Glycoprotein. Amino Acid Squence: TQASSTSFQTKSSEVNLRG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 2943082
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.