Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS PVDF-Plus™ Transfer Membrane

Catalog No. 0973251
Click to view available options
Pore Size:
0.22 μm
0.45 μm

Improves detection limits for more sensitive Western blots.

  • Superior strength: Can withstand aggressive handling or be used with automated equipment without breaking or tearing
  • Low extractables: Ensures tests will be clean with consistent results
  • Exceptional sensitivity: Detects low-level components
  • Hydrophobic:For high protein binding
  • Lot-to-lot consistency: Quality checks ensure consistent binding for dependable results every time
  • BSA protein binding capacity is 125mg/cm2
  • High range of chemical: Resistant to most commonly used chemicals compatible with chemically aggressive solvents

Specifications

Length (Metric) 3 m
Width (Metric) 15 cm
For Use With (Application) Western blots, Immunoblotting, Solid phase assays, Amino acid or protein analyses
Binding Capacity 125 mg/cm2
Color White
Format Roll
Material PVDF
Pore Size 0.45 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.