Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PVR (CD155) Mouse anti-Human, Unconjugated, Clone: SKII.4, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $461.76
Specifications
Antigen | PVR (CD155) |
---|---|
Clone | SKII.4 |
Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF22452MI
|
MilliporeSigma
MABF224525UL |
25 μL |
Each for $182.31
|
|
|||||
MABF22451MI
|
MilliporeSigma
MABF2245100UL |
100 μL |
Each for $461.76
|
|
|||||
Description
PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibits Activity/Function.Specifications
PVR (CD155) | |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Poliovirus receptor;Nectin-like protein 5;NECL-5;CD155 | |
SK-N-S1 human neuroblastoma cells | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
45.31 kDa calculated. |
SKII.4 | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_006496.4 | |
PVR, PVS | |
IgG1 κ | |
Protein G Purified | |
Clone SKII.4 is a mouse monoclonal antibody that specifically detects human PVR (CD155). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title