Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVRIG Antibody (2334B), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB176412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176412 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176412 Supplier Novus Biologicals Supplier No. FAB93652L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PVRIG Monoclonal antibody specifically detects PVRIG in Human samples. It is validated for Flow Cytometry

Specifications

Antigen PVRIG
Applications Flow Cytometry
Classification Monoclonal
Clone 2334B
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias C7orf15MGC138295, MGC104322, MGC138297, MGC2463, poliovirus receptor related immunoglobulin domain containing, Poliovirus receptor-related immunoglobulin domain-containing protein, transmembrane protein PVRIG
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human PVRIG, Thr41-Leu172, Accession # Q6DK17
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 79037
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.