Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXDN Antibody, Novus Biologicals™
SDP

Catalog No. NB096377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096377 Supplier Novus Biologicals Supplier No. NBP305251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PXDN Polyclonal specifically detects PXDN in Human samples. It is validated for Western Blot.

Specifications

Antigen PXDN
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias peroxidasin homolog (Drosophila)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1320-1479 of human PXDN (NP_036425.1). RTRGQFNAFSYHFRGRRSLEFSYQEDKPTKKTRPRKIPSVGRQGEHLSNSTSAFSTRSDASGTNDFREFVLEMQKTITDLRTQIKKLESRLSTTECVDAGGESHANNTKWKKDACTICECKDGQVTCFVEACPPATCAVPVNIPGACCPVCLQKRAEEKP
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7837
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.