Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXK Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP259125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259125 Supplier Novus Biologicals Supplier No. NBP259125

Rabbit Polyclonal Antibody

PXK Polyclonal specifically detects PXK in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen PXK
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias FLJ20335, K-ATPase, K-ATPase long form, Modulator of Na, MONaKA, PX domain containing serine/threonine kinase, PX domain-containing protein kinase-like protein, PX ser/thr kinase v2, PX serine/threonine kinase
Gene Symbols PXK
Host Species Rabbit
Immunogen Maps to a region between 528 and 578
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 54899
Target Species Human
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.