Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXMP4 Antibody (7S41) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP221987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221987 0.1 mg
NB019273 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP221987 Supplier Novus Biologicals Supplier No. NBP221987
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PXMP4 Monoclonal specifically detects PXMP4 in Mouse samples. It is validated for ELISA.

Specifications

Antigen PXMP4
Applications ELISA
Classification Monoclonal
Clone 7S41
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA 1:1000-1:2000
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. P21245
Gene Alias PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa
Gene Symbols PXMP4
Host Species Rat
Immunogen Mouse recombinant PXMP4
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11264
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.