Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PXMP4 Antibody (7S41) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | PXMP4 |
---|---|
Clone | 7S41 |
Concentration | LYOPH |
Applications | ELISA |
Classification | Monoclonal |
Description
PXMP4 Monoclonal specifically detects PXMP4 in Mouse samples. It is validated for ELISA.Specifications
PXMP4 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P21245 | |
11264 | |
IgG2 | |
Protein G purified |
7S41 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa | |
PXMP4 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title