Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXMP4 Antibody (7S41), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB360853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360853 Supplier Novus Biologicals Supplier No. NBP221987MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PXMP4 Monoclonal antibody specifically detects PXMP4 in Mouse samples. It is validated for ELISA

Specifications

Antigen PXMP4
Applications ELISA
Classification Monoclonal
Clone 7S41
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa
Host Species Rat
Immunogen Mouse recombinant PXMP4
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11264
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.