Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXR/NR1I2 Antibody (V11P4G11*E7), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309301 Supplier Novus Biologicals Supplier No. NBP250519MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PXR/NR1I2 Monoclonal antibody specifically detects PXR/NR1I2 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PXR/NR1I2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone V11P4G11*E7
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide EMRSMNEEYT (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8856
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.