Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXR Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$156.65 - $620.58

Specifications

Antigen PXR
Concentration Lot-Specific
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2728
SDP
Encompass
View Documents
Active Motif
39590
10 μL
Each for $156.65
Only null left
Add to Cart
 
50-199-2727
SDP
Encompass
View Documents
Active Motif
39589
200 μL
Each for $620.58
Only null left
Add to Cart
 
Description

Description

This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
Specifications

Specifications

PXR
Western Blot
Unconjugated
Rabbit
Human
O75469
8856
This PXR antibody was raised against a peptide containing amino acid residues 57-71 of human PXR.
Primary
-20°C, Avoid Freeze/Thaw Cycles
NR1I2
Lot-Specific
Polyclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
BXR; mPXR; NR1I2; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; ONR1; Orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR; PAR 1; PAR 2; PAR q; PAR1; PAR2; PARq; pregnane X nuclear receptor variant 2; Pregnane X receptor; pregnane X receptor (nuclear receptor sub family 1, group I, member 2); PRR; Pxr; PXR.1; PXR.2; PXR1; SAR; Steroid and xenobiotic receptor; SXR
NR1I2
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.