Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYK2/FAK2 Antibody, Novus Biologicals™
SDP

Catalog No. NB176656 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176656 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176656 Supplier Novus Biologicals Supplier No. NBP312969
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PYK2/FAK2 Polyclonal antibody specifically detects PYK2/FAK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen PYK2/FAK2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias CADTKCAK-beta, CAK beta, CAKB, Calcium-dependent tyrosine kinase, Cell adhesion kinase beta, EC 2.7.10, FADK2, FAK2EC 2.7.10.2, Focal adhesion kinase 2, PKB, ppyk2, Proline-rich tyrosine kinase 2, protein kinase B, protein tyrosine kinase 2 beta, protein-tyrosine kinase 2-beta, PTK, PTK2B protein tyrosine kinase 2 beta, PYK2Related adhesion focal tyrosine kinase, RAFTKFADK 2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PYK2/FAK2. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cellular Markers, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 2185
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.