Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYK2/FAK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | PYK2/FAK2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PYK2/FAK2 Polyclonal specifically detects PYK2/FAK2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PYK2/FAK2 | |
Polyclonal | |
Rabbit | |
Angiogenesis, Apoptosis, Cancer, Cellular Markers, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
2185 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TLTSPMEYPSPVNSLHTPPLHRHNVFKRHSMREEDFIQPSSREEAQQLWEAEKVKMRQILDKQQKQMVEDYQWLRQEEKSLDPM | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Rat | |
CADTKCAK-beta, CAK beta, CAKB, Calcium-dependent tyrosine kinase, Cell adhesion kinase beta, EC 2.7.10, FADK2, FAK2EC 2.7.10.2, Focal adhesion kinase 2, PKB, Proline-rich tyrosine kinase 2, protein kinase B, protein tyrosine kinase 2 beta, protein-tyrosine kinase 2-beta, PTK, PTK2B protein tyrosine kinase 2 beta, PYK2Related adhesion focal tyrosine kinase, RAFTKFADK 2 | |
PTK2B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title