Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyranose Oxidase Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP17770910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17770910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17770910 Supplier Novus Biologicals Supplier No. NBP177709100UG

Goat Polyclonal Antibody

Pyranose Oxidase Polyclonal specifically detects Pyranose Oxidase in E. coli samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Acetylcholinesterase/ACHE
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Gene Symbols ACHE
Host Species Goat
Immunogen Acetyl Cholinesterase [Bovine Erythrocytes]
Purification Method Delipidation and Defibrination
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 43
Test Specificity Acetylcholine Esterase
Target Species Bovine
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.