Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrethroid hydrolase Ces2e Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB132053 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132053 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132053 Supplier Novus Biologicals Supplier No. NBP306082AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pyrethroid hydrolase Ces2e Polyclonal specifically detects Pyrethroid hydrolase Ces2e in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Pyrethroid hydrolase Ces2e
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Pyrethroid hydrolase Ces2e (NP_766347.1; Met1-His556).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.