Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Closures for Rimless Culture Tubes
SCI_ED

Catalog No. s67136 Shop All Corning Products
Click to view available options
Color:
Natural
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed for culture tubes and flasks with long rimless necks

These Corning polypropylene closures are designed for culture tubes and flasks with long rimless necks. Closure offers two positions: open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination. Closures are autoclavable at 121°C.



Closure ending in -25 fits Cat. No. 4442-125.

Closure ending with -38 fits Cat. Nos. 4442-250, 4442-500, 4442-1 L, and 4442-2L.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Snap Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
For Use With (Equipment) Pyrex™ Flat Bottom Culture Tubes and Flasks with Long Rimless 25 mm O.D. Neck
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.