Learn More
PYREX™ Culture Tubes with Threaded End and Marking Spot Available on GSA/VA Contract for Federal Government customers only.
For tissue culture and general bacteriological work
Manufacturer:  Corning, Inc. 9944716
 View more versions of this product
Catalog No. 14-962B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
15.5mL
16mm
125mm
No
No
White Marking Spot
Round
15-415
Threaded
Without
Borosilicate Glass
Yes
For tissue culture work and general bacteriological use
Bulk
Untreated
Accessories