Learn More
Pyridinium trifluoromethanesulfonate, 97%
Description
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyridinium trifluoromethanesulfonate, esters are used as a catalyst for the reactions of: condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
It is miscible with water, soluble in polar solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 221°C to 223°C |
| Quantity | 5 g |
| Solubility Information | It is miscible with water,soluble in polar solvents. |
| Formula Weight | 229.17 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | Pyridinium trifluoromethanesulfonate |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.