Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Pyridostatin pentahydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Description
Binds and stabilizes G-quadruplexes, inducing DNA damage and cell cycle arrest (Kd = 490 nM); targets the proto-oncogene Src, reducing Src protein abundance and Src-dependent motility in human breast cancer cells. Also targets telomeric G-quadruplexes, inducing telomerase dysfunction. Activates the DNA-dependent protein kinase catalytic sunbunit (DNA-PKcs).
Specifications
Specifications
CAS | 1781882-65-2 |
Quantity | 10 mg |
Molecular Formula | 4-(2-Aminoethoxy)N2,N6-bis[(4-(2-aminoethoxy)-2-quinolinyl]-2,6-pyridinecarboxamide pentahydrochloride |
Product Type | Pyridostatin pentahydrochloride |
Purity | 95% |
Molecular Weight (g/mol) | M.W. (Observed): 778.94 |
Inhibitors | Pyridostatin pentahydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction