Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal Kinase/PDXK Antibody (OTI5H5), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273754FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273754FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273754FR Supplier Novus Biologicals Supplier No. NBP273754FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pyridoxal Kinase/PDXK Monoclonal specifically detects Pyridoxal Kinase/PDXK in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Pyridoxal Kinase/PDXK
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5H5
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias C21orf124, C21orf97, chromosome 21 open reading frame 124, DKFZp566A071, EC 2.7.1.35, FLJ21324, FLJ37311, MGC15873, MGC31754, MGC52346, PKHchromosome 21 open reading frame 97, PNKhuman pyridoxal kinase, EC 2.7.1.3510FLJ31940, PRED79, pyridoxal (pyridoxine, vitamin B6) kinase, pyridoxal kinase, pyridoxamine kinase, Pyridoxine kinase, vitamin B6 kinase
Gene Symbols PDXK
Host Species Mouse
Immunogen Full length recombinant protein of human Pyridoxal Kinase/PDXK (NP_03672) produced in HEK293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8566.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.