Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Pyrimethamine
GREENER_CHOICESDP

Catalog No. 391850 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-185-0 Supplier R&D Systems™ Supplier No. 3918/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent inhibitor of multidrug and toxin extrusion (MATE) transporters; also DHFR and STAT3 inhibitor

Potent inhibitor of multidrug and toxin extrusion (MATE) transporters (Ki values are 46 and 77 nM for human MATE2-K-HEK293 and MATE1-HEK293 cells respectively). Also inhibits dihydrofolate reductase (DHFR) and STAT3. Decreases proliferation of human autosomal dominant polycystic kidney disease cells.

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12H13ClN4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 248.71
Purity >99%
Molecular Weight (g/mol) 248.71
Chemical Name or Material Pyrimethamine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.