Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrogallic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003940 Shop All Avantor Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-940 500 g Wide Mouth Amber Glass Bottle
02-003-939 125 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-003-940 Supplier Avantor Supplier No. 028901
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 500 g
Specific Gravity 1L = 1.45kg
Formula Weight 126.11
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Pyrogallic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.