Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUJIFILM Wako PYROSTAR™ ES-F 80 Test Kit
Shop All FujiFilm Products
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.06 EU/mL
0.125 EU/mL
0.25 EU/mL
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Gel clot sensitivities range from 0.015 to 0.25 EU/mL
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Chances of interference are reduced due to high sensitivity
- Facilitates easier transition from gel clot to KTA

Specifications
Specifications
Form | Lyophilized |
Fill Volume | 2 mL |
Includes | 4 x 2 mL multi-test vials and 1 vial CSE (500 ng/vial) |
Quantity | 80 Tests |
Sensitivity | 0.015 EU/mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction