Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUJIFILM Wako LAL Reagent Water
Click to view available options
Fill Volume:
30 mL
125 mL
Includes:
12 x 125 mL plastic bottles
20 x 30 mL glass bottles
Quantity:
12 Bottles
20 Bottles
Description
- Endotoxin-free water (<0.001 EU/mL)
- Steam-sterilized by USP standards
- Non-LAL reactive
- Great for reconstitution of LAL, RSE and CSE
- Used to perform dilution series of RSE and CSE
- Used for sample dilutions
Specifications
Specifications
| Form | Liquid |
| Fill Volume | 30 mL |
| Includes | 20 x 30 mL glass bottles |
| Quantity | 20 Bottles |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction