Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Carboxylase Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP149536R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149536R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149536R Supplier Novus Biologicals Supplier No. NBP149536R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pyruvate Carboxylase Polyclonal specifically detects Pyruvate Carboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Pyruvate Carboxylase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P11498
Gene Alias EC 6.4.1, EC 6.4.1.1, PCBPyruvic carboxylase, pyruvate carboxylase, pyruvate carboxylase, mitochondrial
Gene Symbols PC
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal region of the human Pyruvate Carboxylase protein (within residues 930-1050). [Swiss-Prot P11498]
Molecular Weight of Antigen 126 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5091
Test Specificity Human and mouse. Immunogen has 96% identity to bovine and porcine. Rat reactivity reported in scientific literature (PMID: 24333689).
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.