Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Dehydrogenase E1-alpha subunit, p Ser293 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11093479T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11093479T 0.025 mL
NB11093479 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11093479T Supplier Novus Biologicals Supplier No. NB11093479SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 43 publications

Pyruvate Dehydrogenase E1-alpha subunit Polyclonal specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoblotting, Knockout Validated.

Specifications

Antigen Pyruvate Dehydrogenase E1-alpha subunit (p Ser293)
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot, Immunoassay
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry reported in scientific literature (PMID 31900478), Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation reported in scientific literature (PMID 27450723), Immunoblotting, Knockout Validated reported in scientific literature (PMID 31742248)
Formulation PBS (pH 7.4) with 0.1% Sodium Azide
Gene Accession No. P08559
Gene Alias EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial
Gene Symbols PDHA1
Host Species Rabbit
Immunogen A synthetic peptide surrounding the phosphorylated serine 293 of the human Pyruvate Dehydrogenase E1-alpha subunit protein. [Swiss-Prot: #P08559]
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Lipid and Metabolism, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5160
Test Specificity This is specific for the phosphorylated Serine 293 form of the PDHE1 alpha protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.