Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Dehydrogenase E1-alpha subunit, p Ser293 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 43 publications

$247.53 - $532.07

Specifications

Antigen Pyruvate Dehydrogenase E1-alpha subunit (p Ser293)
Concentration 1 mg/mL
Dilution Western Blot 1:1000-1:5000, Flow Cytometry reported in scientific literature (PMID 31900478), Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation reported in scientific literature (PMID 27450723), Immunoblotting, Knockout Validated reported in scientific literature (PMID 31742248)
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot, Immunoassay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11093479T
SDP
View Documents
Novus Biologicals
NB11093479SS
0.025 mL
Each for $247.53
Only null left
Add to Cart
 
NB11093479
SDP
View Documents
Novus Biologicals
NB11093479
0.05 mL
Each for $532.07
Only null left
Add to Cart
 
Description

Description

Pyruvate Dehydrogenase E1-alpha subunit Polyclonal specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoblotting, Knockout Validated.
Specifications

Specifications

Pyruvate Dehydrogenase E1-alpha subunit (p Ser293)
Western Blot 1:1000-1:5000, Flow Cytometry reported in scientific literature (PMID 31900478), Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation reported in scientific literature (PMID 27450723), Immunoblotting, Knockout Validated reported in scientific literature (PMID 31742248)
Polyclonal
Rabbit
Cancer, Core ESC Like Genes, Lipid and Metabolism, Signal Transduction, Stem Cell Markers
PBS (pH 7.4) with 0.1% Sodium Azide
EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial
PDHA1
IgG
Affinity Purified
This is specific for the phosphorylated Serine 293 form of the PDHE1 alpha protein.
1 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot, Immunoassay
Unconjugated
RUO
Human, Mouse, Rat
P08559
5160
A synthetic peptide surrounding the phosphorylated serine 293 of the human Pyruvate Dehydrogenase E1-alpha subunit protein. [Swiss-Prot: #P08559]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
43 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.