Learn More
Pyruvate Dehydrogenase E1-alpha subunit, p Ser293 Antibody, DyLight 550, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Pyruvate Dehydrogenase E1-alpha subunit (p Ser293) |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoblot, Immunoassay |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Knockout Validated |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | P08559 |
| Gene Alias | EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial |
| Gene Symbols | PDHA1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.