Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Dehydrogenase E2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB084824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084824 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084824 Supplier Novus Biologicals Supplier No. NBP299340AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pyruvate Dehydrogenase E2 Polyclonal specifically detects Pyruvate Dehydrogenase E2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Pyruvate Dehydrogenase E2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias 70 kDa mitochondrial autoantigen of primary biliary cirrhosis, dihydrolipoamide S-acetyltransferase, DLTA, E2 component of pyruvate dehydrogenase complex, EC 2.3.1, M2 antigen complex 70 kDa subunit, PBC, PDC-E2EC 2.3.1.12, PDCE2mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Pyruvate Dehydrogenase E2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1737
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.