Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Dehydrogenase Kinase 1/PDK1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB090489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090489 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090489 Supplier Novus Biologicals Supplier No. NBP299975G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pyruvate Dehydrogenase Kinase 1/PDK1 Polyclonal specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human samples. It is validated for ELISA.

Specifications

Antigen Pyruvate Dehydrogenase Kinase 1/PDK1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Pyruvate Dehydrogenase Kinase 1/PDK1 (NP_002601.1; Ser29-Ala436).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5163
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.