Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Kinase Antibody, Novus Biologicals™
SDP

Catalog No. NBP244231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244231 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244231 Supplier Novus Biologicals Supplier No. NBP244231
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects Pyruvate Kinase in Rabbit samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting

Pyruvate Kinase Polyclonal specifically detects Pyruvate Kinase in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Pyruvate Kinase
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:4000-1:20000, Immunohistochemistry 1:500-1:2000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P11974
Gene Alias PKM, PKM2
Host Species Goat
Immunogen Pyruvate Kinase (Rabbit Muscle) (Uniprot: P11974)
Molecular Weight of Antigen 58.05 kDa
Purification Method Multi-step
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product has been reported to react with all forms of pyruvate kinase (pan M-PK). Cross-reactivity against pyruvate kinase from other mammalian tissues is expected but has not been specifically determined.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.