Learn More
Pyruvic aldehyde dimethyl acetal, 97+%
Description
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
Solubility
Soluble in water. 350 g/L at 20°C.
Notes
Do not store together with acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | -57°C |
Density | 0.998 |
Boiling Point | 133°C to 135°C |
Flash Point | 34°C (93°F) |
Refractive Index | 1.398 |
Quantity | 500 g |
UN Number | UN1993 |
Beilstein | 1560557 |
Solubility Information | Soluble in water. 350g/L at 20°C. |
Formula Weight | 118.13 |
Show More |
Safety and Handling
Hazard Category | H226-H315-H319 |
Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 228-735-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.