Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Q-VD-OPH Inhibitor Peptide

Applications: Functional, In vitro assay
$404.00 - $1452.00
Specifications
Host Species | Human |
---|---|
Color | Off-white |
For Use With (Application) | Functional, In vitro assay |
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Type | Q-VD-OPH Inhibitor Peptide |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP229391
![]() |
Novus Biologicals™
NBP229391 |
1 mg |
Each for $404.00
|
|
|||||
NBP2293913
![]() |
Novus Biologicals™
NBP2293913MG |
3 mg |
Each for $1,053.00
|
|
|||||
NBP2293915
![]() |
Novus Biologicals™
NBP2293915MG |
5 mg |
Each for $1,452.00
|
|
|||||
Specifications
Human | |
Functional, In vitro assay | |
Q-VD-OPH Inhibitor Peptide | |
Q-VD-OPH |
Off-white | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
513 | |
Semi-solid |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title