Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QA1b Antibody (6A8.6F10.1A6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB22667J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB22667J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB22667J549 Supplier Novus Biologicals Supplier No. NBP226667JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

QA1b Monoclonal specifically detects QA1b in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen QA1b
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 6A8.6F10.1A6
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias H2-Q1, H2-T23
Gene Symbols QA1b
Host Species Mouse
Immunogen Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells.The antibody reacts with the Qa-1b Class Ib molecule.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 15040.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.