Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix TriScreen Positive Control

Catalog No. 23279175 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix Trisceen Control
Packaging:
10 x 1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Packaging
23-279-175 AcroMetrix Trisceen Control 10 x 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-279-175 Supplier Thermo Scientific™ Supplier No. 960811

missing translation for 'validMainframeMsg'

Assess serology assays that test human plasma from donations of whole blood and blood components with these controls, which are for In Vitro Diagnostic Use.

Designed to maintain quality assurance and for the purpose of assessing assay performance of nucleic acid based HIV-1 RNA, HCV RNA, and/or HBV DNA tests. The control product is intended for use with qualitiative assays to provide a means of estimating accuracy, and it has the potential for detecting systematic deviations of nucleic acid test procedures for the qualitative determination of HIV-1 RNA, HCV RNA, and/or HBV DNA. For In Vitro Diagnostic Use.

  • HIV-1 RNA concentration is targeted at 356 copies/mL
  • HCV RNA concentration is targeted at 356 IU/mL
  • HBV DNA concentration is targeted at 356 IU/mL 
TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix Trisceen Control
Detectable Analytes HBV, HIV, HCV
Sample Type Plasma
Type Control
Packaging 10 x 1mL
Quantity 10 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.