Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QIL1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB123537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB123537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB123537 Supplier Novus Biologicals Supplier No. NBP309318100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

QIL1 Polyclonal specifically detects QIL1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen QIL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias C19orf70, chromosome 19 open reading frame 70, P117, Protein P117, protein QIL1, QIL1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of QIL1 (NP_694792). Peptide sequence TGLEMPQLPTPPKIKFPNFRDSWNSGIISVMSALSVAPSKAREYSKEGWE
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125988
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.