Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific QNC-700 Quadra Mini-Vertical PAGE/Blotting System
SCI_ED

Catalog No. p-4085728
Click to view available options
Device Holds:
4- 10 x 10 cm precast/handcast Gel
4-10 x 10 cm precast Gel or 4 blot

Includes

Reservoir with cooling labyrinths and tubing adapters, 2 dual cores, 4 blotting cassettes with sponge pads, and safety cover with attached power leads

Capable of running 4 gels or blots simultaneously at a time.

  • Double-sided cooling labyrinth for temperature-controlled runs
  • Optional self closing quick disconnect valves
  • Fits ClearPAGE™ Precast Gels or glass plate dimensions of either 10 x 10/10 x 9/10 x 8cm (H)

SDS PAGE, Western blotting, acrylamide nucleic acid separation

Specifications

Ambient Temperature (English) 140°F
Ambient Temperature (Metric) 60°C
Certifications/Compliance EN61010-1-1993 (IEC1010-1), CE
Device Holds 4- 10 x 10 cm precast/handcast Gel
Type Quadra Mini Vertical PAGE/Blotting System
Includes 4 Place Gel Wrap™ hand casting bundle (4 combs, 4 sets spacers, 4 Glass plate sets and 4 Gel Wrap™ gaskets, 12 clamps), lower Buffer reservoir with double sided cooling labyrinth and tubing adapters, 2 dual-capacity cor
Volume (English) Buffer 32.12 to 37.19 oz.
Volume (Metric) Buffer 950 to 1100 mL
Dimensions (L x W x H) 17.1 x 17.8 x 15.2 in. (43.4 x 45.2 x 38.6 cm)
Length (English) 17.1 in.
Length (English) Gel 3.93, 3.54, 3.14 in.
Length (Metric) 43.4 cm
Length (Metric) Gel 10, 9, 8 cm
Width (English) 17.8 in.
Width (English) Gel 3.93 in.
Width (Metric) 45.2 cm
Width (Metric) Gel 10 cm
Height (English) 15.2 in.
Height (Metric) 38.6 cm
Voltage 300 V
Wattage 30 w
Amperage 190 mA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.