Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Tocris Bioscience™ QNZ 46
    
    
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                10 mg
                            
                        
                            
                                50 mg
                            
                        
                    Description
GluN2C/GluN2D-selective NMDA receptor non-competitive antagonist (IC50 values are 3, 6, 229, and >300, >300 μM for GluN2D, GluN2C, GluN2A, GluN2B, and GluR1, respectively). Requires binding of glutamate to the GluN2 subunit. 
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
| Quantity | 10 mg | 
| Formula Weight | 443.41 | 
| Purity | >98% | 
| Molecular Weight (g/mol) | 443.415 | 
| Chemical Name or Material | QNZ 46 | 
Safety and Handling
| Recommended Storage | Store at Room Temperature | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction