Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Kraft Specimen Mailers Without Bottles
GSA_VA

With durable metal ringed bottoms and metal screw cap tops

$268.00 - $376.50

Specifications

Applications For mailing liquids
Color Natural
Material Fiberboard
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (English) Exterior Height (English) Exterior Height (Metric) Exterior Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Exterior Height (English) Exterior Height (Metric) Exterior Price Quantity & Availability  
02-991-355
GSA_VA
View Documents
Qorpak™
PAP03116
2.5 in. 6.5 in. 165.1 mm
Case of 24 for $376.50
Only null left
Add to Cart
 
02-991-354
View Documents
Qorpak™
PAP03114
1.75 in. 4.5 in. 114.3 mm
Case of 24 for $272.00
Only null left
Add to Cart
 
02-991-356
View Documents
Qorpak™
PAP03115
2 in. 5.5 in. 139.70 mm
Case of 24 for $268.00
Only null left
Add to Cart
 
Description

Description

  • Meet government regulations for mailing liquids
  • Bottles not included

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

For mailing liquids
Natural
Fiberboard
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.